Diethyl allylphosphonate ≥95.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
D3069-1G
D3069-5G
TCIAD3069-1GEA
206
DKK
TCIAD3069-1G
TCIAD3069-5G
Diethyl allylphosphonate ≥95.0% (by GC)
Diethyl allylphosphonate
Formel:
C₇H₁₅O₃P Kogepunkt: 102°C Flammepunkt: 106°C |
MDL Number:
MFCD00015134 CAS-nummer: 1067-87-4 |
|
Specifikationer Testresultater
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 95.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...