Piperonyl butoxide ≥90.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
Synonymer:
5-[2-(2-Butoxyethoxy)ethoxymethyl]-6-propyl-1,3-benzodioxole, 5-((2-(2-Butoxyethoxy)ethoxy)methyl)-6-propylbenzo[d][1,3]dioxole, NSC 8401
P0458-25G
P0458-500G
TCIAP0458-25GEA
132
DKK
TCIAP0458-25G
TCIAP0458-500G
Piperonyl butoxide ≥90.0% (by GC)
Piperonyl butoxide
Formel:
C₁₉H₃₀O₅ Kogepunkt: 195°C Flammepunkt: 171°C |
MDL Number:
MFCD00005842 CAS-nummer: 51-03-6 EINECS: 200-076-7 UN nummer: 2810 ADR: 6.1,II Merck Index: 14,07476 |
|
Specifikationer Testresultater
Appearance | White to Yellow to Green clear liquid |
Purity (GC) | min. 90.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...