Tetrapropenylsuccinic anhydride (mixture of branched chain isomers) ≥85.0% (by titrimetric analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

T0170-25G T0170-500G
TCIAT0170-25GEA 132 DKK
TCIAT0170-25G TCIAT0170-500G
Tetrapropenylsuccinic anhydride (mixture of branched chain isomers) ≥85.0% (by titrimetric analysis)
Tetrapropenylsuccinic anhydride (mixture of branched chain isomers)
Formel: C₁₆H₂₆O₃
Flammepunkt: 185°C
MDL Number: MFCD00005528
CAS-nummer: 26544-38-7
EINECS: 247-781-6

Bestil nu

Specifikationer Testresultater

Appearance Light yellow to Yellow to Orange clear liquid to cloudy liquid
Purity (Morpholine Method) min. 85.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR