Ammonium 8-anilinonaphthalene-1-sulphonate ≥95.0% (by HPLC, titration analysis) hydrophobic fluorescent probe

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Ammonium 8-(phenylamino)naphthalene-1-sulphonate, 8-Anilino-1-naphthalenesulphonic acid Ammonium salt, Ammonium 8-Anilino-1-naphthalenesulphonate, 8-Anilinonaphthalene-1-sulphonic acid ammonium salt, ANS-NH4

A5351-1G A5351-10G
TCIAA5351-1GEA 140 DKK
TCIAA5351-1G TCIAA5351-10G
Ammonium 8-anilinonaphthalene-1-sulphonate ≥95.0% (by HPLC, titration analysis) hydrophobic fluorescent probe
Ammonium 8-anilinonaphthalene-1-sulphonate
Formel: C₁₆H₁₆N₂O₃S
Smeltepunkt: 237°C
MDL Number: MFCD00012560
CAS-nummer: 28836-03-5
EINECS: 249-265-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=87da2699dc6228db311b038bd54fde416bc7da25&vid=541a5d4b2b8597c951706e7410f946768f022937&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Light yellow to Amber to Dark green powder to crystal
Purity (HPLC) min. 95.0 area%
Purity (Neutralization titration) min. 95.0 %
Solubility in Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR