Sodium-8-anilinonaphthalene-1-sulphonate ≥97.0% (by HPLC, total nitrogen) hydrophobic fluorescent probe

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: ANS-Na, 8-Anilino-1-naphthalenesulphonic acid sodium salt, Sodium 8-Anilino-1-naphthalenesulphonate

A5352-1G A5352-5G
TCIAA5352-1GEA 575 DKK
TCIAA5352-1G TCIAA5352-5G
Sodium-8-anilinonaphthalene-1-sulphonate ≥97.0% (by HPLC, total nitrogen) hydrophobic fluorescent probe
Sodium-8-anilinonaphthalene-1-sulphonate
Formel: C₁₆H₁₂NNaO₃S
MDL Number: MFCD00054279
CAS-nummer: 1445-19-8
EINECS: 215-890-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=87be8c122ab9522b7e7526df4ea372827db811f1&vid=bf551a3e57a8a0a0f1cb9a2a780c646b34cf8d4b&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Amber to Dark green powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (with Total Nitrogen) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR