6-Acetoxy-2-naphthoic acid ≥98.0% (by HPLC, titration analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
6-Acetoxy-2-naphthoic Acid
A0970-1G
A0970-5G
TCIAA0970-1GEA
403
DKK
TCIAA0970-1G
TCIAA0970-5G
6-Acetoxy-2-naphthoic acid ≥98.0% (by HPLC, titration analysis)
6-Acetoxy-2-naphthoic acid
Formel:
C₁₃H₁₀O₄ Molekylvægt: 230.22 g/mol |
MDL Number:
MFCD00059556 CAS-nummer: 17295-26-0 |
|
Specifikationer Testresultater
Appearance | White to Light yellow powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 225.0 to 230.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...