1-Bromo-2-naphthoic acid ≥98.0% (by GC, titration analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
B1617-1G
B1617-100MG
B1617-5G
TCIAB1617-1GEA
870
DKK
TCIAB1617-1G
TCIAB1617-100MG
TCIAB1617-5G
1-Bromo-2-naphthoic acid ≥98.0% (by GC, titration analysis)
1-Bromo-2-naphthoic acid
Formel:
C₁₁H₇BrO₂ |
MDL Number:
MFCD00021408 CAS-nummer: 20717-79-7 EINECS: 243-984-9 |
|
Specifikationer Testresultater
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 187.0 to 191.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...