Dichlorotitanium diisopropoxide ≥95.0% (by titrimetric analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Dichloro(diisopropoxy)titanium

D2257-25G D2257-5G
TCIAD2257-25GEA 1800 DKK
TCIAD2257-25G TCIAD2257-5G
Dichlorotitanium diisopropoxide ≥95.0% (by titrimetric analysis)
Dichlorotitanium diisopropoxide
Formel: C₆H₁₄Cl₂O₂Ti
Opbevaringstemperatur: Fryser
MDL Number: MFCD00191416
CAS-nummer: 762-99-2
EINECS: 212-106-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=66d382986ee3bad51077247543cc2ead8a7179d7&vid=66d382986ee3bad51077247543cc2ead8a7179d7&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Light yellow powder to crystal
Purity (Chelometric Titration) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR