Diethyl (4-fluorobenzyl)phosphonate ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: (4-Fluorobenzyl)phosphonic acid diethyl ester, Diethyl 4-fluorobenzylphosphonate

D3324-25G D3324-5G
TCIAD3324-25GEA 1850 DKK
TCIAD3324-25G TCIAD3324-5G
Diethyl (4-fluorobenzyl)phosphonate ≥98.0% (by GC)
Diethyl (4-fluorobenzyl)phosphonate
Formel: C₁₁H₁₆FO₃P
Molekylvægt: 246.2200 g/mol
Kogepunkt: 150°C
MDL Number: MFCD06795966
CAS-nummer: 63909-58-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=557c3a63e0551cd21892fa7d3431b9fc5cd6d332&vid=557c3a63e0551cd21892fa7d3431b9fc5cd6d332&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR