4-Amino-3-chloro-5-nitrobenzotrifluoride ≥97%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: 2-Chloro-6-nitro-4-(trifluoromethyl)aniline

B20938.03 B20938.06
B20938.03EA 232 DKK
B20938.03 B20938.06
4-Amino-3-chloro-5-nitrobenzotrifluoride ≥97%
4-Amino-3-chloro-5-nitrobenzotrifluoride

Purity: 97% CAS No.: 57729-79-0 Molecular Formula: C7H4ClF3N2O2 Formula Weight: 240.58 Melting Point: 63-67° UN No.: 2811 MDL No.: MFCD00042153

Formel: C₇H₄ClF₃N₂O₂
Molekylvægt: 240.58 g/mol
Smeltepunkt: 63…67 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00042153
CAS-nummer: 57729-79-0
UN nummer: 2811
ADR: 6.1 III

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR