(3-Bromo-4-methoxyphenyl)acetonitrile ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: 2-(3-Bromo-4-methoxyphenyl)acetonitrile

B1455-25G B1455-5G
TCIAB1455-25GEA 1470 DKK
TCIAB1455-25G TCIAB1455-5G
(3-Bromo-4-methoxyphenyl)acetonitrile ≥98.0% (by GC)
(3-Bromo-4-methoxyphenyl)acetonitrile
Formel: C₉H₈BrNO
Kogepunkt: 178°C
MDL Number: MFCD00016391
CAS-nummer: 772-59-8
UN nummer: 2811
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6113e03d3300497d88c27d4898ea76e86c68d033&vid=6113e03d3300497d88c27d4898ea76e86c68d033&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Purity (GC) min. 98.0 %
Melting point 54.0 to 57.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR