4-Methoxy-β-nitrostyrene ≥98%
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
A12745.06
A12745.14
A12745.22
A12745.06EA
994
DKK
A12745.06
A12745.14
A12745.22
4-Methoxy-β-nitrostyrene ≥98%
4-Methoxy-β-nitrostyrene
Purity: 98%
CAS No.: 5576-97-6
Molecular Formula: C9H9NO3
Formula Weight: 179.18
Storage Temperature: 2°C to 8°C
Melting Point: 86-88°
Beilstein Reference: 2047935
MDL No.: MFCD00024826
Formel:
C₉H₉NO₃ Smeltepunkt: 86…88 °C |
MDL Number:
MFCD00024826 CAS-nummer: 3179-10-0 |