Progesterone ≥98.0% (by HPLC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
P0478-25G
P0478-5G
TCIAP0478-25GEA
1060
DKK
TCIAP0478-25G
TCIAP0478-5G
Progesterone ≥98.0% (by HPLC)
Progesterone
Formel:
C₂₁H₃₀O₂ |
MDL Number:
MFCD00003658 CAS-nummer: 57-83-0 EINECS: 200-350-6 Merck Index: 14,07773 |
|
Specifikationer Testresultater
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (UV-vis method) | 97.0 to 103.0 % |
Melting point | 128.0 to 133.0 deg-C |
Specific rotation [a]20/D | +179.0 to +185.0 deg (C=2, Dioxane) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...