Tetraethyl ethylenebisphosphonate ≥98.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
Tetraethyl ethylenediphosphonate, Tetraethyl ethane-1,2-diylbis(phosphonate)
T2294-5G
T2294-1G
TCIAT2294-5GEA
673
DKK
TCIAT2294-5G
TCIAT2294-1G
Tetraethyl ethylenebisphosphonate ≥98.0% (by GC)
Tetraethyl ethylenebisphosphonate
Formel:
C₁₀H₂₄O₆P₂ Molekylvægt: 302.25 g/mol Kogepunkt: 202°C |
MDL Number:
MFCD00015133 CAS-nummer: 995-32-4 EINECS: 213-625-0 |
|
Specifikationer Testresultater
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...