Diethyl (hydroxymethyl)phosphonate ≥97.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

D3813-100G D3813-25G
TCIAD3813-100GEA 559 DKK
TCIAD3813-100G TCIAD3813-25G
Diethyl (hydroxymethyl)phosphonate ≥97.0% (by GC)
Diethyl (hydroxymethyl)phosphonate
Formel: C₅H₁₃O₄P
Molekylvægt: 168.13 g/mol
Kogepunkt: 105°C
Opbevaringstemperatur: Køleskab
MDL Number: MFCD00014418
CAS-nummer: 3084-40-0
EINECS: 221-391-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=79ca1d24e570b32cce9aec35a8c5d01df056f8de&vid=79ca1d24e570b32cce9aec35a8c5d01df056f8de&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR