2-Naphthylacetonitrile ≥97%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

L09384.06 L09384.14
L09384.06EA 819 DKK
L09384.06 L09384.14
2-Naphthylacetonitrile ≥97%
2-Naphthylacetonitrile

Purity: 97% CAS No.: 7498-57-9 Molecular Formula: C12H9N Formula Weight: 167.21 Melting Point: 82-84° Boiling Point: 303° UN No.: 3439 MDL No.: MFCD00004122

Formel: C₁₂H₉N
Molekylvægt: 167.21 g/mol
Kogepunkt: 303 °C
Smeltepunkt: 82…84 °C
Massefylde: 1.092
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00004122
CAS-nummer: 7498-57-9
EINECS: 231-352-5
UN nummer: 3439
ADR: 6.1 III

Bestil nu

Specifikationer

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR