Methyl (diethylphosphono)acetate ≥97%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Diethyl methoxycarbonylmethanephosphonate, Methyl 2-(diethoxyphosphoryl)acetate, Methyl diethylphosphonoacetate

A10644.14 A10644.22 A10644.36
A10644.14EA 717 DKK
A10644.14 A10644.22 A10644.36
Methyl (diethylphosphono)acetate ≥97%
Methyl (diethylphosphono)acetate

Purity: 97% CAS No.: 1067-74-9 Molecular Formula: C7H15O5P Formula Weight: 210.17 Flash Point: 110°(230°F) Refractive Index: 1.4320 Boiling Point: 126-128°/8mm Beilstein Reference: 1782397 MDL No.: MFCD00009081

Formel: C₇H₁₅O₅P
Kogepunkt: 126…128 °C (8 mmHg)
Massefylde: 1,145
Flammepunkt: >110 °C (230 °F)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00009081
CAS-nummer: 1067-74-9
EINECS: 213-938-2

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR