Triethyl-2-phosphonopropionate ≥98.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Synonymer:
Diethyl [1-(ethoxycarbonyl)ethyl]phosphonate, Ethyl 2-(diethoxyphosphoryl)propanoate
T2135-25G
T2135-5G
TCIAT2135-25GEA
928
DKK
TCIAT2135-25G
TCIAT2135-5G
Triethyl-2-phosphonopropionate ≥98.0% (by GC)
Triethyl-2-phosphonopropionate
Formel:
C₉H₁₉O₅P Molekylvægt: 238.22 g/mol Kogepunkt: 144°C Flammepunkt: 88°C |
MDL Number:
MFCD00009159 CAS-nummer: 3699-66-9 EINECS: 223-033-4 |
|
Specifikationer Testresultater
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...