tert-Butyl (diethylphosphono)acetate ≥97.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: tert-Butyl 2-(diethoxyphosphoryl)acetate, Diethyl(2-tert-butoxycarbonylmethyl)phosphonate

B2814-1G B2814-5G
TCIAB2814-1GEA 189 DKK
TCIAB2814-1G TCIAB2814-5G
tert-Butyl (diethylphosphono)acetate ≥97.0% (by GC)
tert-Butyl (diethylphosphono)acetate
Formel: C₁₀H₂₁O₅P
Kogepunkt: 152°C
MDL Number: MFCD00075414
CAS-nummer: 27784-76-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bea5fb6ec8766dcde1519032f136581a40108093&vid=bea5fb6ec8766dcde1519032f136581a40108093&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR