2-Hydroxy-2-methylpropiophenone ≥96.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 2-Benzoyl-2-propanol, 2-Hydroxy-2-methyl-1-phenylpropan-1-one

H0991-25G H0991-500G H0991-100G
TCIAH0991-25GEA 148 DKK
TCIAH0991-25G TCIAH0991-500G TCIAH0991-100G
2-Hydroxy-2-methylpropiophenone ≥96.0% (by GC)
2-Hydroxy-2-methylpropiophenone
Formel: C₁₀H₁₂O₂
Kogepunkt: 110°C
MDL Number: MFCD00040710
CAS-nummer: 7473-98-5
EINECS: 231-272-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e85ae67e8470380cfe86a69a7d05530785c1fed2&vid=e85ae67e8470380cfe86a69a7d05530785c1fed2&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR