4,4'-Dihydroxydiphenylmethane ≥98%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 4,4'-Methylenediphenol

A11417.03 A11417.06
A11417.03EA 407 DKK
A11417.03 A11417.06
4,4'-Dihydroxydiphenylmethane ≥98%
4,4'-Dihydroxydiphenylmethane

Purity: 98% CAS No.: 620-92-8 Molecular Formula: C13H12O2 Formula Weight: 200.24 Melting Point: 159-163° Beilstein Reference: 2049425

Formel: C₁₃H₁₂O₂
Molekylvægt: 200.24 g/mol
Smeltepunkt: 159…163 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00002385
CAS-nummer: 620-92-8
EINECS: 210-658-2

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR