3,4-Dimethoxyphenylacetone ≥97%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: 1-(3,4-Dimethoxyphenyl)propan-2-one

L05477.09 L05477.18
L05477.09EA 322 DKK
L05477.09 L05477.18
3,4-Dimethoxyphenylacetone ≥97%
3,4-Dimethoxyphenylacetone

Purity: 97% CAS No.: 776-99-8 Molecular Formula: C11H14O3 Formula Weight: 194.23 Flash Point: 110°(230°F) Refractive Index: 1.5358 Boiling Point: 154-158°/12mm Beilstein Reference: 1107410 MDL No.: MFCD00008772

Formel: C₁₁H₁₄O₃
Kogepunkt: 154…158 °C (12 mmHg)
Massefylde: 1,115
Flammepunkt: >110 °C (230 °F)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00008772
CAS-nummer: 776-99-8
EINECS: 212-285-0

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR