Traumatic acid (trans-2-dodecene-1,12-dioic acid)
Leverandør: Cayman Chemical
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Synonymer: trans-2-Dodecene-1,12-dioic acid, Traumatic acid
88820-500 88820-1 88820-5 88820-10
CAYM88820-500EA 1860 DKK
CAYM88820-500 CAYM88820-1 CAYM88820-5 CAYM88820-10
Traumatic acid (trans-2-dodecene-1,12-dioic acid)
Traumatic acid (trans-2-dodecene-1,12-dioic acid)
Formel: C₁₂H₂₀O₄
MDL Number: MFCD00004443
CAS-nummer: 6402-36-4
EINECS: 229-019-4