4,4'-(Hexafluoroisopropylidene)diphenol ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 4,4′-(Hexafluoroisopropylidene)diphenol, 2,2-Bis(4-hydroxyphenyl)hexafluoropropane, 4,4'-(Perfluoropropane-2,2-diyl)diphenol

B0945-25G B0945-5G B0945-100G
TCIAB0945-25GEA 247 DKK
TCIAB0945-25G TCIAB0945-5G TCIAB0945-100G
4,4'-(Hexafluoroisopropylidene)diphenol ≥98.0% (by GC)
4,4'-(Hexafluoroisopropylidene)diphenol
Formel: C₁₅H₁₀F₆O₂
Molekylvægt: 336.24 g/mol
MDL Number: MFCD00000439
CAS-nummer: 1478-61-1
EINECS: 216-036-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8d72a5b8ecfdf9913fb0284faeda3c3f0ac7929f&vid=8d72a5b8ecfdf9913fb0284faeda3c3f0ac7929f&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Gray to Red powder to crystal
Purity (GC) min. 98.0 %
Melting point 159.0 to 163.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR