4-Amino-2,3,5,6-tetrafluorobenzamide ≥98.0% (by GC, total nitrogen)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

A0949-1G A0949-5G
TCIAA0949-1GEA 279 DKK
TCIAA0949-1G TCIAA0949-5G
4-Amino-2,3,5,6-tetrafluorobenzamide ≥98.0% (by GC, total nitrogen)
4-Amino-2,3,5,6-tetrafluorobenzamide
Formel: C₇H₄F₄N₂O
MDL Number: MFCD00008000
CAS-nummer: 1548-74-9
EINECS: 216-285-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ae8d5337e19149cf9284e71a25914e098e2d8e01&vid=ae8d5337e19149cf9284e71a25914e098e2d8e01&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Purity (with Total Nitrogen) min. 98.0 %
Melting point 183.0 to 188.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR