2-Phenylethanethiol ≥97.0% (by GC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Phenethyl mercaptan

P1715-25G P1715-5G
TCIAP1715-25GEA 682 DKK
TCIAP1715-25G TCIAP1715-5G
2-Phenylethanethiol ≥97.0% (by GC, titration analysis)
2-Phenylethanethiol
Formel: C₈H₁₀S
Kogepunkt: 218°C
Flammepunkt: 90°C
MDL Number: MFCD00004891
CAS-nummer: 4410-99-5
EINECS: 224-563-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7469a08f418cf23c0e0e2d9d48aa0dfd8d0b3477&vid=7469a08f418cf23c0e0e2d9d48aa0dfd8d0b3477&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 97.0 %
Purity (Iodometric Titration) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR