Carmine 40,0% (by HPLC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: CI 75470

C0543-25G C0543-5G
TCIAC0543-25GEA 879 DKK
TCIAC0543-25G TCIAC0543-5G
Carmine 40,0% (by HPLC)
Carmine
Formel: C₂₂H₂₀O₁₃
Molekylvægt: 492.389 g/mol
MDL Number: MFCD00167028
CAS-nummer: 1390-65-4
EINECS: 215-724-4
Merck Index: 14,01843
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b3f32e3e6d99d5fce16b0d6307d12ad004a79421&vid=b3f32e3e6d99d5fce16b0d6307d12ad004a79421&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Red to Dark red to Brown powder to crystal
Purity (HPLC) min. 40.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR