Phenylglyoxylic acid ≥97.0% (by GC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

B1129-25G B1129-5G
TCIAB1129-25GEA 1270 DKK
TCIAB1129-25G TCIAB1129-5G
Phenylglyoxylic acid ≥97.0% (by GC, titration analysis)
Phenylglyoxylic acid
Formel: C₈H₆O₃
Kogepunkt: 138°C
MDL Number: MFCD00002575
CAS-nummer: 611-73-4
EINECS: 210-278-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8e4e872b1a5e601f55677e62acb2d3896ecdc6eb&vid=8e4e872b1a5e601f55677e62acb2d3896ecdc6eb&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Light yellow powder to crystal
Purity (GC) min. 97.0 %
Purity (Neutralization titration) min. 97.0 %
Melting point 61.0 to 67.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR