4-Methoxybenzhydrol ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

M1278-5G M1278-25G
TCIAM1278-5GEA 230 DKK
TCIAM1278-5G TCIAM1278-25G
4-Methoxybenzhydrol ≥98.0% (by GC)
4-Methoxybenzhydrol
Formel: C₁₄H₁₄O₂
Molekylvægt: 214.26 g/mol
MDL Number: MFCD00014398
CAS-nummer: 720-44-5
EINECS: 211-953-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=922da9d393bd0073b18ca1ec5d2bb9c3263103c5&vid=922da9d393bd0073b18ca1ec5d2bb9c3263103c5&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 63.0 to 67.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR