2-tert-Butylpyrazine ≥97.0% (by GC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

B3394-1G B3394-5G
TCIAB3394-1GEA 476 DKK
TCIAB3394-1G TCIAB3394-5G
2-tert-Butylpyrazine ≥97.0% (by GC, titration analysis)
2-tert-Butylpyrazine
Formel: C₈H₁₂N₂
MDL Number: MFCD00069193
CAS-nummer: 32741-11-0
EINECS: 251-186-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e3e48c5fc47ebca883145d4c21916baa731689f5&vid=e3e48c5fc47ebca883145d4c21916baa731689f5&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 97.0 %
Purity (Nonaqueous Titration) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR