(1-Naphthylmethyl)triphenylphosphonium chloride ≥98.0% (by titrimetric analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: (Naphthalen-1-ylmethyl)triphenylphosphonium chloride

N0700-25G N0700-5G
TCIAN0700-25GEA 1830 DKK
TCIAN0700-25G TCIAN0700-5G
(1-Naphthylmethyl)triphenylphosphonium chloride ≥98.0% (by titrimetric analysis)
(1-Naphthylmethyl)triphenylphosphonium chloride
Formel: C₂₉H₂₄ClP
MDL Number: MFCD00031658
CAS-nummer: 23277-00-1
EINECS: 245-548-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=76fb6c840efc47fbd7524a1d90c83c83175cf54a&vid=76fb6c840efc47fbd7524a1d90c83c83175cf54a&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR