4-(Dimethylamino)cinnamaldehyde ≥98.0% (by titrimetric analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
p-Dimethylamino-Cinnamaldehyde
D0648-25G
D0648-5G
TCIAD0648-25GEA
1470
DKK
TCIAD0648-25G
TCIAD0648-5G
4-(Dimethylamino)cinnamaldehyde ≥98.0% (by titrimetric analysis)
4-(Dimethylamino)cinnamaldehyde
Formel:
C₁₁H₁₃NO Molekylvægt: 175.23 g/mol |
MDL Number:
MFCD00007002 CAS-nummer: 6203-18-5 EINECS: 228-267-0 |
|
Specifikationer Testresultater
Appearance | White to Brown powder to crystal |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 139.0 to 142.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...