4-Chlorotetrahydropyran ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

C2273-25G C2273-5G
TCIAC2273-25GEA 2480 DKK
TCIAC2273-25G TCIAC2273-5G
4-Chlorotetrahydropyran ≥98.0% (by GC)
4-Chlorotetrahydropyran
Formel: C₅H₉ClO
Kogepunkt: 150°C
Flammepunkt: 46°C
MDL Number: MFCD00074911
CAS-nummer: 1768-64-5
UN nummer: 1993
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bc4b67ec972bcc7ef2394334323ad5fb27f31941&vid=bc4b67ec972bcc7ef2394334323ad5fb27f31941&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR