tert-Butyl-N-hydroxycarbamate ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

H0848-25G H0848-5G
TCIAH0848-25GEA 673 DKK
TCIAH0848-25G TCIAH0848-5G
tert-Butyl-N-hydroxycarbamate ≥98.0% (by GC)
tert-Butyl-N-hydroxycarbamate
Formel: C₅H₁₁NO₃
MDL Number: MFCD00002107
CAS-nummer: 36016-38-3
EINECS: 252-836-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=284c8edfda65b224aabb573f43efc8795f3ab4b1&vid=284c8edfda65b224aabb573f43efc8795f3ab4b1&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 55.0 to 59.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR