Allyl acetoacetate ≥95.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: Allyl Acetoacetate, Acetoacetic acid allyl ester

A1981-25G A1981-500G
TCIAA1981-25GEA 288 DKK
TCIAA1981-25G TCIAA1981-500G
Allyl acetoacetate ≥95.0% (by GC)
Allyl acetoacetate
Formel: C₇H₁₀O₃
Kogepunkt: 195°C
Flammepunkt: 76°C
MDL Number: MFCD00009811
CAS-nummer: 1118-84-9
EINECS: 214-269-9
UN nummer: 2810
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=93da40a8f76995e39d0c0dcf15bb6e9c0bce61e9&vid=93da40a8f76995e39d0c0dcf15bb6e9c0bce61e9&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR