2,3-Dihydrobenzo[b]furan ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

D1583-25G D1583-5G
TCIAD1583-25GEA 977 DKK
TCIAD1583-25G TCIAD1583-5G
2,3-Dihydrobenzo[b]furan ≥98.0% (by GC)
2,3-Dihydrobenzo[b]furan
Formel: C₈H₈O
Kogepunkt: 189°C
Flammepunkt: 66°C
MDL Number: MFCD00005855
CAS-nummer: 496-16-2
EINECS: 207-817-3
Merck Index: 14,02559
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=74fd5b8fcf6853454bf981e3272f29fa29f150e4&vid=74fd5b8fcf6853454bf981e3272f29fa29f150e4&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Yellow to Green clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR