Azidotrimethylsilane ≥95.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
T0801-25G
T0801-5G
T0801-100G
TCIAT0801-25GEA
542
DKK
TCIAT0801-25G
TCIAT0801-5G
TCIAT0801-100G
Azidotrimethylsilane ≥95.0% (by GC)
Azidotrimethylsilane
Formel:
C₃H₉N₃Si Kogepunkt: 97°C Opbevaringstemperatur: Køleskab |
MDL Number:
MFCD00001986 CAS-nummer: 4648-54-8 EINECS: 225-078-5 UN nummer: 1992 ADR: 3,II |
|
Specifikationer Testresultater
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 95.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...