4-Chloro-6-methoxypyrimidine ≥98.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
C2369-1G
C2369-5G
TCIAC2369-1GEA
411
DKK
TCIAC2369-1G
TCIAC2369-5G
4-Chloro-6-methoxypyrimidine ≥98.0% (by GC)
4-Chloro-6-methoxypyrimidine
Formel:
C₅H₅ClN₂O Kogepunkt: 80°C |
MDL Number:
MFCD06200211 CAS-nummer: 26452-81-3 |
|
Specifikationer Testresultater
Appearance | White or Colorless to Light yellow powder to lump to clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...