(Aminomethyl)phosphonic acid ≥97.0% (by titrimetric analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: (Aminomethyl)phosphonic Acid, AMPA

A1072-1G A1072-5G
TCIAA1072-1GEA 1090 DKK
TCIAA1072-1G TCIAA1072-5G
(Aminomethyl)phosphonic acid ≥97.0% (by titrimetric analysis)
(Aminomethyl)phosphonic acid
Formel: CH₆NO₃P
MDL Number: MFCD00008105
CAS-nummer: 1066-51-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6dc38bdb8bdbe69069afce3e98993dff9a9ff2bc&vid=6dc38bdb8bdbe69069afce3e98993dff9a9ff2bc&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystaline
Purity (Neutralization titration) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR