Methyl-4-chloroacetoacetate ≥96.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: Methyl 4-chloro-3-oxobutanoate

C0910-25ML C0910-250ML
TCIAC0910-25MLEA 279 DKK
TCIAC0910-25ML TCIAC0910-250ML
Methyl-4-chloroacetoacetate ≥96.0% (by GC)
Methyl-4-chloroacetoacetate
Formel: C₅H₇ClO₃
Molekylvægt: 150.56 g/mol
Kogepunkt: 97°C
Flammepunkt: 108°C
MDL Number: MFCD00000938
CAS-nummer: 32807-28-6
EINECS: 251-230-5
UN nummer: 2922
ADR: 8 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5e0afb4022d45b3122f014060a99a08687f30ec2&vid=5e0afb4022d45b3122f014060a99a08687f30ec2&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR