(Chloromethyl)triphenylphosphonium chloride ≥98.0% (by HPLC, titration analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
(chloromethyl)triphenylphosphoniumchloride
C1009-25G
C1009-5G
TCIAC1009-25GEA
1460
DKK
TCIAC1009-25G
TCIAC1009-5G
(Chloromethyl)triphenylphosphonium chloride ≥98.0% (by HPLC, titration analysis)
(Chloromethyl)triphenylphosphonium chloride
Formel:
C₁₉H₁₇Cl₂P |
MDL Number:
MFCD00011867 CAS-nummer: 5293-84-5 EINECS: 226-139-9 |
|
Specifikationer Testresultater
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Argentmetric Titration) | min. 98.0 % |
Solubility in Water | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...