5-Amino-2-chlorophenol ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 2-Chloro-5-aminophenol

A1976-25G A1976-5G
TCIAA1976-25GEA 1570 DKK
TCIAA1976-25G TCIAA1976-5G
5-Amino-2-chlorophenol ≥98.0% (by GC, titration analysis)
5-Amino-2-chlorophenol
Formel: C₆H₆ClNO
Molekylvægt: 143.57 g/mol
Smeltepunkt: 160°C
MDL Number: MFCD01707858
CAS-nummer: 6358-06-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=58850dbadf541e9303459ddf3da889366adcd2fb&vid=58850dbadf541e9303459ddf3da889366adcd2fb&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Gray to Brown powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR