4-Nitroquinoline-N-oxide ≥98.0% (by HPLC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: 4-Nitroquinoline 1-oxide, 4-Nitroquinoline N-oxide

N0250-1G N0250-5G
TCIAN0250-1GEA 329 DKK
TCIAN0250-1G TCIAN0250-5G
4-Nitroquinoline-N-oxide ≥98.0% (by HPLC, titration analysis)
4-Nitroquinoline-N-oxide
Formel: C₉H₆N₂O₃
Molekylvægt: 190.1555 g/mol
MDL Number: MFCD00006738
CAS-nummer: 56-57-5
EINECS: 200-281-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=09ce7fb423ee109eb32349fe82806b04e748ef16&vid=09ce7fb423ee109eb32349fe82806b04e748ef16&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Light yellow to Brown powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Solubility in hot Acetone very faint turbidity


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR