N,N'-Diphenylbenzidine ≥98.0% (by HPLC, total nitrogen)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
4,4'-Bis(phenylamino)biphenyl, N4,N4'-Diphenyl-[1,1'-biphenyl]-4,4'-diamine
D0878-25G
D0878-5G
TCIAD0878-25GEA
1320
DKK
TCIAD0878-25G
TCIAD0878-5G
N,N'-Diphenylbenzidine ≥98.0% (by HPLC, total nitrogen)
N,N'-Diphenylbenzidine
Formel:
C₂₄H₂₀N₂ |
MDL Number:
MFCD00003016 CAS-nummer: 531-91-9 EINECS: 208-521-7 Merck Index: 14,03319 |
|
Specifikationer Testresultater
Appearance | White to Gray to Brown powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Solubility in hot THF | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...