Erythromycin ≥98.0% (by titrimetric analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

E0751-5G E0751-25G
TCIAE0751-5GEA 312 DKK
TCIAE0751-5G TCIAE0751-25G
Erythromycin ≥98.0% (by titrimetric analysis)
Erythromycin
Formel: C₃₇H₆₇NO₁₃
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00084654
CAS-nummer: 114-07-8
EINECS: 204-040-1
Merck Index: 14,03681
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=86ea48766b0d44e8cda19e67545245daada50047&vid=86ea48766b0d44e8cda19e67545245daada50047&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Orange to Green powder to crystal
Purity (Nonaqueous Titration) min. 98.0 % (calcd.on anh.substance)
Specific rotation [a]20/D -71.0 to -78.0 deg (C=2, EtOH) (calcd.on anh.substance)
Water max. 10.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR