6-(Dimethylamino)purine ≥98.0% (by HPLC, titration analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
D3894-1G
D3894-5G
TCIAD3894-1GEA
600
DKK
TCIAD3894-1G
TCIAD3894-5G
6-(Dimethylamino)purine ≥98.0% (by HPLC, titration analysis)
6-(Dimethylamino)purine
Formel:
C₇H₉N₅ Smeltepunkt: 260°C |
MDL Number:
MFCD00005573 CAS-nummer: 938-55-6 EINECS: 213-344-3 |
|
Specifikationer Testresultater
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...