Diphenylphosphinic acid ≥98.0% (by HPLC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

D3627-25G D3627-5G
TCIAD3627-25GEA 665 DKK
TCIAD3627-25G TCIAD3627-5G
Diphenylphosphinic acid ≥98.0% (by HPLC, titration analysis)
Diphenylphosphinic acid
Formel: C₁₂H₁₁O₂P
MDL Number: MFCD00002132
CAS-nummer: 1707-03-5
EINECS: 216-948-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=520fe85103f5f028adc108b240844345ae886dd8&vid=520fe85103f5f028adc108b240844345ae886dd8&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 193.0 to 197.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR