N,N'-Bis(3-aminopropyl)ethylenediamine ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: 1,5,8,12-Tetraazadodecane

B1952-25ML B1952-500ML
TCIAB1952-25MLEA 132 DKK
TCIAB1952-25ML TCIAB1952-500ML
N,N'-Bis(3-aminopropyl)ethylenediamine ≥98.0% (by GC, titration analysis)
N,N'-Bis(3-aminopropyl)ethylenediamine
Formel: C₈H₂₂N₄
Kogepunkt: 165°C
Flammepunkt: 142°C
MDL Number: MFCD00008210
CAS-nummer: 10563-26-5
EINECS: 234-147-9
UN nummer: 2927
ADR: 6.1,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a4650cb4293343c75c6a1aa312ee32924382358e&vid=a4650cb4293343c75c6a1aa312ee32924382358e&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR