9-Bromo-1-nonanol ≥95.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 9-Bromononan-1-ol

B1785-25G B1785-5G
TCIAB1785-25GEA 772 DKK
TCIAB1785-25G TCIAB1785-5G
9-Bromo-1-nonanol ≥95.0% (by GC)
9-Bromo-1-nonanol
Formel: C₉H₁₉BrO
Molekylvægt: 223.15 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD00063348
CAS-nummer: 55362-80-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d9e46b101f879743046f9a5271e5a076d482df6d&vid=d9e46b101f879743046f9a5271e5a076d482df6d&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Light yellow to Light orange powder to lump
Purity (GC) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR