7-Bromo-1-heptanol ≥88.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
B1852-1G
B1852-5G
TCIAB1852-1GEA
279
DKK
TCIAB1852-1G
TCIAB1852-5G
7-Bromo-1-heptanol ≥88.0% (by GC)
7-Bromo-1-heptanol
Formel:
C₇H₁₅BrO Opbevaringstemperatur: Fryser |
MDL Number:
MFCD00041708 CAS-nummer: 10160-24-4 |
|
Specifikationer Testresultater
Purity (GC) | min. 88.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...